missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ LB Medium, Large Capsules
Culture media used for the growth of bacteria.
Brand: MP Biomedicals™ 113002046
Additional Details : Weight : 0.01000kg
Description
- Formulations for the E. coli growth media contain 10g of tryptone and 5g of yeast extract per liter, 10g NaCl per liter
Specifications
Molecular Biology Certified™ | |
Capsule | |
Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L | |
5000 g |
15°C to 30°C | |
Microbiology Media | |
For growth of bacteria |