Learn More
Thermo Scientific™ LanthaScreen™ Thiol Reactive Tb Chelate
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
Description
- As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Thiol Reactive Tb Chelate labeling reagent is available for assay development
- The thiol-reactive maleimide group can be conjugated to virtually any peptide or protein that contains one or more accessible thiol moieties, such as the thiol group found on a cysteine residue
- Energy transfer from the terbium donor to a suitable acceptor such as fluorescein is readily detected by monitoring an increase in acceptor fluorescence intensity
- See the user guide for an application-based protocol for cysteine-containing peptides or proteins
Specifications
Specifications
| Quantity | 10 μg |
| Product Type | Thiol Reactive Tb Chelate |
| Content And Storage | LanthaScreen™ TR-FRET Labeling Reagents are provided lyophilized. Store at -20°C, desiccated, and protected from light. After reconstitution, terbium lanthanide chelates should be stored on ice and used within the same day. |
| Shipping Condition | Dry Ice |
| Product Line | LanthaScreen |