Learn More
Invitrogen™ LanthaScreen™ Tb-labeled Anti-Rabbit Antibody Kit
Product Code. 10216262
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
1mg
25µg
Product Code. 10216262
Brand: Invitrogen™ PV3775
To facilitate in-house assay development around multiple target classes and assay formats, Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents.
To facilitate in-house assay development around multiple target classes and assay formats, Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents. Tb-labeled secondary antibodies bind to their corresponding species IgG with low nanomolar apparent Kd's, and show negligible cross reactivity with IgG's from other species. Each Tb-labeled anti-species antibody is provided with a corresponding fluorescein-labeled IgG as a positive control.
In a kinase assay utilizing the Tb-labeled antibody generic reagent, anti-phosphopeptide primary antibody is not labeled and the detection step is performed by the addition of the Tb-labeled secondary antibody. This approach can be applied to other target classes and assay formats.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer (-5°C to -30°C).
Quantity 1 mg
Reactivity Rabbit
Shipping Condition Dry Ice
Conjugate Lanthanide Chelates, Terbium (Tb)
Product Line LanthaScreen