Learn More
Labconco™ Sample Blocks for RapidVap™ N2/48 Evaporation Systems
27746.47 NOK - 28157.06 NOK
Requires: Glassware
| Product Code | Brand | Holds | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Holds | Price | Quantity & Availability | |||||
|
11746419
|
Labconco™
7482400 |
35 mL Tubes |
33405.00 NOK 27746.47 NOK / Pack of 1 Save 5658.53 NOK 16% Off |
Estimated Shipment: 11-12-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
11726649
|
Labconco™
7482200 |
10 mL Tubes |
32440.00 NOK 28157.06 NOK / Pack of 1 Save 4282.94 NOK 13% Off |
Estimated Shipment: 20-01-2027
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
- Four different interchangeable aluminum accessory blocks, coated with PTFE
- Sample blocks hold either 48 smaller (4.5 mL to 26 mL) or eight larger (450 mL) samples
- Glassware for all blocks sold separately
†For tubes up to maximum capacity shown. Actual sample volume is 66–75% total tube capacity and depends on tube shape, height and vortex speed.