Learn More
Heathrow Scientific™ Lab Wipe Holder
GREENER_CHOICE
Product Code. 12377619 Shop All Fisherbrand Products
Change view
Click to view available options
Mounting:
Double-sided Tape
Screw Mount
Dimensions (L x W x H):
118 x 122 x 84 mm
320 x 127 x 97 mm
Size:
Small
Large
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mounting Dimensions (L x W x H) Size unitSize
12377619 Double-sided Tape 118 x 122 x 84 mm Small Each
12387619 Screw Mount 320 x 127 x 97 mm Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12377619
  • ONLINE EXCLUSIVE PRICE
    200.00 NOK
    185.58 NOK / Pack of 1
    Save 14.42 NOK
    7% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
Holds lab wipe boxes such as Kimwipes™ and other major brands. Lab Wipe Holder can be mounted anywhere lab wipes are needed.
  • Clear acrylic holders fit boxes of small or large lab wipes.
  • Mount on a shelf or wall.
  • The smaller version attaches with double-stick tape and the larger version with screws.
  • Mounting materials included.
Specifications
Product Type Labwipe Holder
Color Clear
Dimensions (L x W x H) 118 x 122 x 84 mm
Holds Labwipes
Material Acrylic
Mounting Double-sided Tape
No. of Compartments 1
Product Style Side Loading
Includes Mounting materials
Size Small