Learn More
Fisherbrand™ Lab Drying Rack Replacement Basket
Product Code. 17130041 Shop All Fisherbrand Products
Click to view available options
Holds:
Lab Glassware
Unit Size:
Pack of 2
Product Code. 17130041
Only null left
Add to basket
Add to basket
Lab Drying Rack Replacement Basket for Fisherbrand™ Lab Drying Rack Set
  • Drying Basket to keep small items and slotted to hold pipettes safe while drying
Specifications
Autoclavable Not Autoclavable
Color Clear
Holds Lab Glassware
Material ABS
Positions 1
Shape Square
Sterility Non-sterile
For Use With (Equipment) Lab Drying Rack (120791 F)
Size 5.6 x 2.7 x 2.0 in.
Dimensions (L x W x H) 142.24 x 68.58 x 50.8 mm
Product Type Replacement Basket
Shelves None
Height (Metric) 5.1 cm
Width (Metric) 6.9 cm