missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | L3MBTL3 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17501366
|
Proteintech
28112-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17591356
|
Proteintech
28112-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The L3MBTL3 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human L3MBTL3. This antibody recognizes human antigen. The L3MBTL3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| L3MBTL3 | |
| Polyclonal | |
| Rabbit | |
| 84456 | |
| Primary |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Human | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title