Learn More
L3MBTL3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28112-1-AP-150UL
 View more versions of this product
Product Code. 17591356
  • 4836.10 NOK / 150µL
Description
The L3MBTL3 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human L3MBTL3. This antibody recognizes human antigen. The L3MBTL3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
L3MBTL3
Polyclonal
Rabbit
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
150 μL
84456