Learn More
L3MBTL3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28112-1-AP-20UL
Additional Details : Weight : 0.00020kg
Description
The L3MBTL3 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human L3MBTL3. This antibody recognizes human antigen. The L3MBTL3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
L3MBTL3 | |
Polyclonal | |
Rabbit | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
20 μL | |
84456 | |
IgG |