Learn More
L-Plastin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55208-1-AP-150UL
 View more versions of this product
Product Code. 16876695
  • 4836.10 NOK / 150µL
Description
L-plastin is a leukocyte specific actin bundling protein from fimbrin family of proteins. It has two actin-binding domains and two EF hand type calcium-binding domains. It gets phosphorylated on the serine residue in its headpiece region in response to a variety of inflammatory stimuli. It is hypothesized that L-plastin may play a role in regulating integrin mediated adhesion in leukocytes. L-plastin is expressed in majority of human cancer cell lines derived from solid tumors. Its expression is positively upregulated by testosteronse in andrgen receptor positive prostrate and breast cancer cells through three androgen receptor binding elements located in the upstream region of the gene.
Specifications
Specifications
L-Plastin
Polyclonal
Unconjugated
LCP1
L PLASTIN, LCP 1, LCP1, LPL, Lymphocyte cytosolic protein 1, Plastin 2, PLS2
Rabbit
Antigen Affinity Chromatography
RUO
18826, 3936
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P13796, Q61233
Lcp1
Peptide
150 μL
Primary
Human, Mouse