missing translation for 'onlineSavingsMsg'
Learn More

KL Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen KL
Concentration 233 μg/mL
Applications Western Blot, Immunoprecipitation, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16845995
View Documents
Proteintech
28100-1-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16835995
View Documents
Proteintech
28100-1-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The KL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KL. This antibody recognizes human, mouse antigen. The KL antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.

Specifications

Specifications

KL
Western Blot, Immunoprecipitation, Immunohistochemistry
Unconjugated
Human, Mouse
IgG
233 μg/mL
Polyclonal
Rabbit
9365
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.