Learn More
KL Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28100-1-AP-150UL
 View more versions of this product
Product Code. 16835995
  • 4836.10 NOK / 150µL
Description
The KL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KL. This antibody recognizes human, mouse antigen. The KL antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
Specifications
KL
Polyclonal
Unconjugated
150 μL
9365
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry
233 μg/mL
Rabbit
Primary
Human, Mouse