Learn More
KL Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28100-1-AP-150UL
This item is not returnable.
View return policy
Description
The KL antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KL. This antibody recognizes human, mouse antigen. The KL antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
| KL | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 9365 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry | |
| 233 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |