missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kindlin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | Kindlin 2 |
|---|---|
| Concentration | 0.36 mg/mL |
| Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16848543
|
Proteintech
11453-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16838543
|
Proteintech
11453-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
11453-1-AP is a rabbit polyclonal antibody against human FERMT2.
This gene encodes a myosin IIA heavy chain that contains an IQ domain and a myosin head-like domain. The protein is involved in several important functions, including cytokinesis, cell motility and maintece of cell shape. Defects in MYH9 are the cause of non-syndromic sensorineural deafness autosomal domit type 17, Epstein syndrome, Alport syndrome with macrothrombocytopenia, Sebastian syndrome, Fechtner syndrome and macrothrombocytopenia with progressive sensorineural deafness.Specifications
| Kindlin 2 | |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q8CIB5, Q96AC1 | |
| 10979, 218952, 289992 | |
| Kindlin 2 Fusion Protein Ag2013 | |
| Primary | |
| -20°C | |
| FERMT2 |
| 0.36 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Fermitin family homolog 2, FERMT2, KIND2, Kindlin 2, mig 2, MIG2, PLEKHC1, UNC112, UNC112B | |
| Fermt2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title