missing translation for 'onlineSavingsMsg'
Learn More

Kindlin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11453-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16848543

  • 1415.13 NOK / 20µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

11453-1-AP is a rabbit polyclonal antibody against human FERMT2.

This gene encodes a myosin IIA heavy chain that contains an IQ domain and a myosin head-like domain. The protein is involved in several important functions, including cytokinesis, cell motility and maintece of cell shape. Defects in MYH9 are the cause of non-syndromic sensorineural deafness autosomal domit type 17, Epstein syndrome, Alport syndrome with macrothrombocytopenia, Sebastian syndrome, Fechtner syndrome and macrothrombocytopenia with progressive sensorineural deafness.
Specifications

Specifications

Kindlin 2
Polyclonal
Unconjugated
FERMT2
Fermitin family homolog 2, FERMT2, KIND2, Kindlin 2, mig 2, MIG2, PLEKHC1, UNC112, UNC112B
Rabbit
Antigen Affinity Chromatography
RUO
10979, 218952, 289992
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry
0.36 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8CIB5, Q96AC1
Fermt2
Kindlin 2 Fusion Protein Ag2013
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers