Learn More
DWK Life Sciences Kimble™ KIMAX™ 5-Place NMR Tube Washers
Description
These complete five-place washers use flexible PTFE tubing and a vacuum source (not included) to direct a power stream of wash solvent to thoroughly clean the insides of 7 and 8 in. NMR tubes.
- Two sizes available for 3 and 5mm diameter tubes
- Both sizes come with a 1L reservoir bottle
- All wetted parts are constructed of chemical-resistant PTFE or borosilicate glass
- An O-ring slips over each tube to form a vacuum-tight seal before tubes are positioned for cleaning
- With a simple twist, the wash valve can be configured to wash and then dry
- Cycles can be repeated, or cleaned tubes can be removed immediately for use
Specifications
Specifications
| Material | PTFE or Borosilicate Glass |
| Product Type | NMR Tube Washer |
| For Use With (Equipment) | 5 mm dia. Tubes |