Learn More
Unity Lab Services™ KI Test for Biosafety Cabinet (≥ 1.8 m2)
Click to view available options
Duration:
1 Day
Unit Size:
Each
Description
- Equipment is required to pass EN12469 certification prior to this test
- Required for UK
- This is add-on to STARTBSC[PF] for new equipment, and an add on to BSCCERT for existing equipment
- Includes travel and labor unless extensive travel is needed
- All additional works, as i.e. waiting time or correctional works, will be invoiced additionally according to our recent valid service pricelist
Specifications
Specifications
| For Use With (Equipment) | Biosafety Cabinets |
| Certifications/Compliance | BS EN 12469 (Europe) or NSF 49 (US) |
| Duration | 1 Day |
| Product Type | European KI Test |
| Product Line | Thermo Scientific™ |
| Service Offering | Performance Test |