Learn More
Unity Lab Services™ KI Test for Biosafety Cabinet (≤ 1.5 m2)
Product Code. 17879602
Change view
Click to view available options
Duration:
1 Day
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Duration Unit Size
17879602 1 Day Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Strongly recommended (and sometimes required) in addition to basic certification (part no BSCCERT)
Product Code. 17879602 Supplier Unity Lab Services™ Supplier No. BSCKI15S
 Restricted Item
Estimated Shipment: 15-09-2026
Strongly recommended (and sometimes required) in addition to basic certification (part no BSCCERT)
European KI test for biosafety cabinets 1.5 mt or narrower; Equipment is required to pass certification prior to this service; Required for UK. This is add-on to STARTBSC[PF]
  • Equipment is required to pass EN12469 certification prior to this test
  • Required for UK
  • This is add-on to STARTBSC[PF] for new equipment, and an add on to BSCCERT for existing equipment
  • Includes travel and labor unless extensive travel is needed
  • All additional works, as i.e. waiting time or correctional works, will be invoiced additionally according to our recent valid service pricelist
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Biosafety Cabinets
Certifications/Compliance BS EN 12469 (Europe) or NSF 49 (US)
Duration 1 Day
Product Type European KI Test
Product Line Thermo Scientific
Service Offering Performance Test