missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KHSRP (Human) Recombinant Protein
Human KHSRP partial ORF ( NP_003676, 151 a.a. - 239 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00008570-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: VPDGMVGLIIGRGGEQINKIQQDSGCKVQISPDSGGLPERSVSLTGAPESVQKAKMMLDDIVSRGRGGPPGQFHDNANGGQNGTVQEIM
Specifications
NP_003676 | |
KH-type splicing regulatory protein | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KHSRP | |
GST |
8570 | |
Wheat germ expression system | |
10 μg | |
FBP2, FUBP2, KSRP, MGC99676 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |