missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KHSRP (Human) Recombinant Protein
Human KHSRP partial ORF ( NP_003676, 151 a.a. - 239 a.a.) recombinant protein with GST-tag at N-terminal.
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_003676 |
---|---|
Gene ID (Entrez) | 8570 |
Name | KH-type splicing regulatory protein |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138855
|
Abnova™
H00008570-Q01.10UG |
10 μg |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16148855
|
Abnova™
H00008570-Q01.25UG |
25 μg |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: VPDGMVGLIIGRGGEQINKIQQDSGCKVQISPDSGGLPERSVSLTGAPESVQKAKMMLDDIVSRGRGGPPGQFHDNANGGQNGTVQEIM
Specifications
NP_003676 | |
KH-type splicing regulatory protein | |
125% SDS-PAGE Stained with Coomassie Blue | |
FBP2, FUBP2, KSRP, MGC99676 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
8570 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KHSRP | |
GST |