Learn More
KD Scientific Tygon™ E-Food Tubing For Allegro Peristaltic Pumps
Non-toxic clear tubing designed for food products
Brand:  KD Scientific 785621
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15500540
  • 2320.00 NOK / Each
Estimated Shipment: 12-07-2024
Description
  • Made of thermoplastic, PVC based firm transparent material with plasticizer
  • Smooth bore tubing available in various sizes
Application Compatibility
  • Viscous fluids: Excellent
  • Acid and alkalies: Good
  • Sterile fluids: Fair
  • Organic solvents: No
Specifications
Specifications
19/100 in.
50 ft.
L/S 25
4.8 mm
1520 cm