Learn More
KCNMB2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16606-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16886264
  • 4593.76 NOK / 150µL
Estimated Shipment: 08-05-2024
Description
The KCNMB2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KCNMB2. This antibody recognizes human antigen. The KCNMB2 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
KCNMB2
Polyclonal
Unconjugated
150 μL
10242
IgG
Immunohistochemistry, ELISA
247 μg/mL