missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNMB2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | KCNMB2 |
---|---|
Concentration | 247 μg/mL |
Applications | Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896264
|
Proteintech
16606-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16886264
|
Proteintech
16606-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The KCNMB2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KCNMB2. This antibody recognizes human antigen. The KCNMB2 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
KCNMB2 | |
Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG |
247 μg/mL | |
Polyclonal | |
Rabbit | |
10242 | |
Primary |