Learn More
KCNMB2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16606-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16896264
  • 1611.84 NOK / 20µL
Estimated Shipment: 23-05-2024
Description
The KCNMB2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KCNMB2. This antibody recognizes human antigen. The KCNMB2 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
KCNMB2
Polyclonal
Unconjugated
20 μL
10242
IgG
Immunohistochemistry, ELISA
247 μg/mL