Learn More
Mettler Toledo™ KCl 3M Electrode Filling Solution
Product Code. 11932535
Change view
Click to view available options
Quantity:
25 mL
250 mL
6 x 250 mL
Unit Size:
1500mL
250mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11932535 6 x 250 mL 1500mL
11907781 25 mL 25mL
10780412 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11932535 Supplier Mettler Toledo™ Supplier No. 51350080
  • ONLINE EXCLUSIVE PRICE
    1265.00 NOK
    1214.92 NOK / 1500mL
    Save 50.08 NOK
    3% Off
View all promotions
 Restricted Item
Estimated Shipment: 15-12-2026
Equipped with a special cap to facilitate easy filling of electrodes. Mettler Toledo™ KCl 3M Electrode Filling Solution is an electrolyte solution that enables optimum performance of pH and ion-selective electrodes. This bridge electrolyte contains no silver ions than can contaminate samples.
An electrode is the main actor of pH and ion measurement. They require special care and handling. The electrode should remain filled with fresh, clean electrolyte solution. Regularly refilling or replacing the electrolyte solution is necessary to maintain good electrode performance.
The reference electrode of a pH or Ion-Selective Electrode (ISE) should be filled only with the recommended electrolyte solution. The correct electrolyte minimizes junction potentials and provides optimum temperature and time response.
3mol/L Potassium Chloride solution is appropriate for most applications.
  • Used by most Mettler Toledo pH sensors, due to the ARGENTHAL™ reference system
  • DX-series ISE half-cells can also use a 3M KCl electrolyte bridge solution
  • No risk of contaminating samples with silver ions
  • Equipped with a special cap to facilitate easy filling of electrodes
  • Available in 25mL and 250mL bottles, and a 6 × 250mL bulk pack
TRUSTED_SUSTAINABILITY
Specifications
Color Colorless
Concentration 3 mol/L KCl
For Use With (Equipment) ARGENTHAL™ Reference System pH Electrodes, DX Series ISE Half Cells
NIST Traceable Yes
Packaging Bottle with Special Filling Cap
pH Range Neutral
Quantity 6 x 250 mL