Learn More
Cole-Parmer™ Jenway™ Epoxy Bodied Conductivity Cell
Product Code. 13107354
Change view
Click to view available options
Type:
Conductivity Cell
Unit Size:
Each
Product Code. Type unitSize
13107354 Conductivity Cell Each
1 options
Product Code. 13107354
Brand: Cole-Parmer™ 9995970
Only null left
Specifications
Cell Constant 1
For Use With (Equipment) The Enterprise 470 Unit
Type Conductivity Cell
Material Epoxy