missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12972-1-AP-20UL
This item is not returnable.
View return policy
Description
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. It acts as an adhesive ligand for interacting with a variety of immune cell types and may play a role in lymphocyte homing to secondary lymphoid organs.Specifications
| JAM2 | |
| Polyclonal | |
| Unconjugated | |
| JAM2 | |
| C21orf43, CD322, JAM 2, JAM B, JAM2, JAMB, junctional adhesion molecule 2, Junctional adhesion molecule B, PRO245, VE JAM, VEJAM | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 58494, 619374, 67374 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P57087, Q9JI59 | |
| Jam2 | |
| JAM2 Fusion Protein Ag3431 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction