Learn More
ITGB1BP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21359-1-AP-20UL
This item is not returnable.
View return policy
Description
ITGB1BP2 may play a role during maturation and/or organization of muscles cells.Specifications
| ITGB1BP2 | |
| Polyclonal | |
| Unconjugated | |
| ITGB1BP2 | |
| CHORDC3, ITGB1BP, ITGB1BP2, MELUSIN, MSTP015 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26548, 26549, 317258 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9R000, Q9UKP3 | |
| Itgb1bp2 | |
| ITGB1BP2 Fusion Protein Ag15808 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |