missing translation for 'onlineSavingsMsg'
Learn More

ITGB1BP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen ITGB1BP2
Concentration 0.18 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16850965
View Documents
Proteintech
21359-1-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16840965
View Documents
Proteintech
21359-1-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ITGB1BP2 may play a role during maturation and/or organization of muscles cells.
Specifications

Specifications

ITGB1BP2
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Rat, Mouse, Human
Q9R000, Q9UKP3
26548, 26549, 317258
ITGB1BP2 Fusion Protein Ag15808
Primary
-20°C
ITGB1BP2
0.18 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CHORDC3, ITGB1BP, ITGB1BP2, MELUSIN, MSTP015
Itgb1bp2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.