Learn More
IRS1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
Antigen IRS1
Concentration 0.3 mg/mL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16837414
View Documents
Proteintech
17509-1-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
16827414
View Documents
Proteintech
17509-1-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Description
Description
XPOT is a protein belonging to the RAN-GTPase exportin family that mediates export of tRNA from the nucleus to the cytoplasm. Translocation of tRNA to the cytoplasm occurs once exportin has bound both tRNA and GTP-bound RAN.
Specifications
Specifications
IRS1
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Human
P35568
3667
IRS1 Fusion Protein Ag11714
Primary
-20°C
IRS1
0.3 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
HIRS 1, insulin receptor substrate 1, IRS 1, IRS1
IRS1
IgG
Antigen Affinity Chromatography