Learn More
IRS1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17509-1-AP-20UL
This item is not returnable.
View return policy
Description
XPOT is a protein belonging to the RAN-GTPase exportin family that mediates export of tRNA from the nucleus to the cytoplasm. Translocation of tRNA to the cytoplasm occurs once exportin has bound both tRNA and GTP-bound RAN.Specifications
| IRS1 | |
| Polyclonal | |
| Unconjugated | |
| IRS1 | |
| HIRS 1, insulin receptor substrate 1, IRS 1, IRS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3667 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35568 | |
| IRS1 | |
| IRS1 Fusion Protein Ag11714 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |