Learn More
Memmert™ Iq/Oq Document With Device-Specific Works Test Data (1 Temperature Value), For Ich750
Product Code. 16326697 Shop All Memmert Products
Product Code. 16326697
Brand: Memmert™ ICH750D00127
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes temperature distribution survey for one free-selectable temperature value.
  • IQ/OQ document with works test data for chamber
  • 9 measuring points (size 30), 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand), PQ check list as support for validation by customer
Specifications
Product Type IQ/OQ Documentation