Learn More
Memmert™ Iq/Oq Document W/Device-Specific Works Test Data (Temperature And Humidity Value), For Hcp150
Shop All Memmert ProductsDescription
Includes
Works test data, temperature distribution survey
- IQ/OQ document with works test data for chamber
- 27 measuring points to DIN 12880:2007-05, PQ check list as support for validation by customer (models HPP and ICH)
Specifications
Specifications
| For Use With (Equipment) | 110, 260, 750 Models Constant Climate Chamber |
| Product Type | IQ/OQ Documentation |
| For Use With (Application) | One Free-Selectable Temperature and Humidity Value for 27 Measuring Points to DIN 12880:2007-05 |