Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - CO2 and Temperature
Product Code. 17149994 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
ICO105
ICO150
ICO240
ICO50
Unit Size:
Each
Product Code. 17149994
Brand: Memmert™ ICO240D38898
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For one free-selectable CO2 and temperature value
  • Includes temperature distribution survey at Memmert
  • For 27 measuring points to DIN 12880:2007-05
  • PQ check list as support for validation by customer.
Specifications
For Use With (Equipment) ICO240
Product Type IQ/OQ Document with Device-specific Works Test Data