Learn More
Memmert™ IQ Document with Test Data for UN110
Product Code. 17579755 Shop All Memmert Products
Click to view available options
:
For Use With: UN110
Product Code. 17579755
Brand: Memmert™ UN110D00124
IQ document with device-specific works test data for validation by customer.
  • OQ/PQ check list as support for validation by customer
Specifications
For Use With (Equipment) UN110 Oven
Product Type IQ Document
Includes Test Data