Learn More
Memmert™ IQ Document with Test Data for UF450
Product Code. 17579335 Shop All Memmert Products
Click to view available options
:
For Use With: UF450
Product Code. 17579335
Brand: Memmert™ UF450D00124
IQ document with device-specific works test data for validation by customer.
  • OQ/PQ check list as support for validation by customer
Specifications
For Use With (Equipment) UF450 Oven
Product Type IQ Document
Includes Test Data