Learn More
IPO7 Mouse anti-Human, Unconjugated, Clone: 1E7C12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67525-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16850206
  • 3828.53 NOK / 150µL
Estimated Shipment: 10-10-2023
Description
The IPO7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IPO7. This antibody recognizes human, mouse, rat antigen. The IPO7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
IPO7
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, ELISA
1E7C12
Unconjugated
150 μL
10527
IgG1