missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IPO7 Mouse anti-Human, Unconjugated, Clone: 1E7C12, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | IPO7 |
|---|---|
| Clone | 1E7C12 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16860206
|
Proteintech
67525-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16850206
|
Proteintech
67525-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The IPO7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IPO7. This antibody recognizes human, mouse, rat antigen. The IPO7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| IPO7 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 10527 | |
| Primary |
| 1E7C12 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title