Learn More
SI Analytics™ IonLine pH Combination Electrode
Product Code. 10523524 Shop All SI Analytics Products
Click to view available options
Length (Metric) Cable:
1 m
Unit Size:
Each
Product Code. 10523524
Brand: SI Analytics™ 285114300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
SI Analytics™ IonLine pH Combination Electrode is a micro-electrode with ID function, temperature sensor, and a fixed cable.
  • Temp. sensor Pt 1000
  • 1 m cable with DIN and banana plug
  • Glass shaft
  • Pt diaphragm
  • KCl 3 mol/l
  • Iodine/Iodide-ref. system
  • Cylindrical membrane
TRUSTED_SUSTAINABILITY
Specifications
Material A-Glass
pH Range 0 to 14
Temperature (Metric) Operating -5°C to +100°C
Type Micro-Electrode
Length (Metric) 40 to 130 mm
Length (Metric) Cable 1 m
Diameter (Metric) Body 12 mm
Diameter (Metric) Cap 5 mm sensor