Learn More
Invitrogen™ UltraPure™ DEPC-Treated Water
Product Code. 11588636
Change view
Click to view available options
Quantity:
4 x 1.25 mL
1 L
4 x 100 mL
Unit Size:
1 L
400mL
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11588636 4 x 1.25 mL Pack of 4
11528616 4 x 100 mL 400mL
10636503 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11588636 Supplier Invitrogen™ Supplier No. 10813012
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
Autoclaved to remove the DEPC and 0.1-μm membrane-filtered.
UltraPure™ DEPC-treated Water is suitable for use with RNA. It is prepared by incubating with 0.1% diethylpyrocarbonate (DEPC), and is then autoclaved to remove the DEPC. Sterile filtered.
Performance and Quality Testing: No DNase or RNase activity detected.
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
pH 6 to 8
Grade Molecular Biology
Packaging Tube
Treatment(s) DEPC-Treated
Quantity 4 x 1.25 mL
Purification Method Autoclaved, Membrane-Filtered
Shipping Condition Approved for shipment on Wet or Dry Ice
Product Line UltraPure