Learn More
Invitrogen™ S. cerevisiae Hansen BY4730
Product Code. 10149252
Change view
Click to view available options
:
S. cerevisiae Hansen BY4730
Product Code.
10149252 S. cerevisiae Hansen BY4730
1 options
Product Code. 10149252
Brand: Invitrogen 95400.BY4730
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For cloning and transformation
The S. cerevisiae Hansen strains are the parental yeast strains from which deletions were derived.
Order Info
Shipping Condition: Room Temperature