missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-ROX, SE (5-Carboxy-X-Rhodamine, Succinimidyl Ester), single isomer

Product Code. 11590146
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11590146 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11590146 Supplier Invitrogen™ Supplier No. C6125
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

5-ROX, SE (5-Carboxy-X-Rhodamine, Succinimidyl Ester), single isomer

The isomeric 5-ROX, SE is an amine-reactive form of carboxy-X-rhodamine and is widely used for oligonucleotide labeling and automated DNA sequencing applications.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type 5-ROX SE
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye ROX
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.