Learn More
Miele™ Insert Half Segment
Product Code. 13182280 Shop All Miele Products
Change view
Click to view available options
Depth (Metric):
445 mm
Unit Size:
Each
Product Code. Depth (Metric) unitSize
13182280 445 mm Each
1 options
Product Code. 13182280
Brand: Miele™ 3808600
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Insert, Half
For Use With (Equipment) 210 Slides, for upper basket, lower basket
Depth (Metric) 445 mm
Height (Metric) 73 mm
Width (Metric) 200 mm
Quantity 1 Each