Learn More
Fisherbrand™ Insert For Maxi Boxes
Shop All Fisherbrand Products
Click to view available options
Tube Size:
8 x 1.5mL
15 x 5mL
8 x 20mL
8 x 50mL
15 x 15mL
Unit Size:
Each
Description
- Inserts for Maxi Boxes
- Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
- Also supplied is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids
- These serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
- Both the Beta and Gamma storage bins are available in five sizes, while the two largest bin models have wheels for easy transportation
- Optional heavy duty drawstring bags may also be purchased
Specifications
Specifications
| For Use With (Equipment) | For Maxi Boxes |
| Height (Metric) | 40mm |
| Product Type | Insert |
| Tube Size | 15 x 5mL |
| Width (Metric) | 160mm |