Learn More
Elma™ Insert Covers
Product Code. 17891357
Change view
Click to view available options
Model:
100/120
150/180/300
180
30/40
60
80
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
17891357 150/180/300 Each
17881357 30/40 Each
17831297 60 Each
17861297 80 Each
17811367 100/120 Each
17831347 180 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17891357 Supplier Elma™ Supplier No. 1110260
  • ONLINE EXCLUSIVE PRICE
    755.00 NOK
    561.07 NOK / Pack of 1
    Save 193.93 NOK
    25% Off
Estimated Shipment: 16-12-2026
Insert cover contains two rubber rings and is designed for hanging cleaning cups or glasses in the cleaning basket.
  • Elma ultrasonic units can be operated with or without a cover
  • For more efficient operation, Elma offers various insert covers made of robust and easy-to-clean stainless steel
  • Covers make it possible to clean objects made of different materials simultaneously – but separately
  • If used correctly, all Elma inserts will help to prevent damage to the ultrasonic tank and thus extend the service life of the ultrasonic cleaner
  • Allows cleaning with aggressive fluids
  • Prolongs the service life of the unit
Specifications
Product Type Lid
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic Med, Elmasonic P, Elmasonic S, Elmasonic Select, Elmasonic EASY
Material Stainless Steel
Model 150/180/300