Description
- Holding frames for securing inserts E 106 to E 111
- Enables E 135 insert for baby bottles to be loaded in an E 555/2
- Enables E 135 insert for baby bottles to be loaded in an E 735/2
Specifications
Specifications
| Material | Stainless Steel |
| Color | Stainless Steel |
| For Use With (Equipment) | Washer-disinfectors, laboratory |