Learn More
Miele Professional™ Insert
Insert E 500 for the optimum loading of 8 solder frames
Brand: Miele Professional™ 5376820
Additional Details : Weight : 3.50000kg
Description
Equipped with 8 compartments, spacing 53 mmSpecifications
Stainless Steel | |
Stainless Steel | |
8 Printed circuit boards | |
Washer-disinfectors, laboratory |