Learn More
Miele™ Insert 1/2
Shop All Miele Products
Click to view available options
Product Type:
Insert, Half
Unit Size:
Each
Specifications
Specifications
| Product Type | Insert, Half |
| Model | E 106/1 |
| For Use With (Equipment) | Wide neck flasks, measuring cylinders etc. |
| Includes | 26 small spring hooks x 105mm height (distance beween spring hooks is approx. 60mm) |
| Depth (Metric) | 410 mm |
| Height (Metric) | 116 mm |
| Width (Metric) | 195 mm |
| Quantity | 1 Each |