Learn More
Miele™ Insert 1/2
Product Code. 10292831 Shop All Miele Products
Click to view available options
Product Type:
Insert, Half
Unit Size:
Each
Product Code. 10292831
Brand: Miele™ 3808420
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Insert, Half
Model E 111
For Use With (Equipment) 8 Beakers, 600 to 1000mL
Includes 8 x 3 stay bars
Depth (Metric) 460 mm
Height (Metric) 205 mm
Width (Metric) 230 mm
Quantity 1 Each