Learn More
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection
Shop All Mettler Toledo Products
Click to view available options
Connector Types:
S7-BNC
S7-BNC, IP67 waterproof
S7-DIN 19262
S7-Lemo 00
S7-None
Length (Metric) Cable:
1.2 m
10 m
3.0 m
5.0 m
Unit Size:
Each
Description
- Detachable
- Resuable
Specifications
Specifications
| Connector Types | S7-DIN 19262 |
| For Use With (Equipment) | Mettler Toledo InLab Sensors |
| For Use With (Application) | To connect Mettler Toledo InLab sensors to any meter |
| Length (English) Cable | 9.8 ft. |
| Length (Metric) Cable | 3.0 m |