Learn More
Mettler Toledo™ InLab™ Sensor Cables: MultiPin™ Connection
Product Code. 15503181
Change view
Click to view available options
Connector Types:
MultiPin-BNC/1 x 4mm banana
MultiPin-BNC/RCA (cinch)
MultiPin-BNC/RCA (cinch), IP67 waterproof
MultiPin-DIN 19262/1 x 4mm banana
MultiPin-DIN/RCA (cinch)
MultiPin-Lemo 00/2 x 4mm banana
Length (Metric) Cable:
1.2 m
1.8 m
3.0 m
5.0 m
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Length (Metric) Cable Unit Size
15503181 MultiPin-BNC/RCA (cinch) 1.2 m Each
15529651 MultiPin-DIN/RCA (cinch) 1.2 m Each
15509651 MultiPin-BNC/RCA (cinch) 5.0 m Each
15599641 MultiPin-BNC/RCA (cinch) 3.0 m Each
15519651 MultiPin-BNC/1 x 4mm banana 1.2 m Each
15559651 MultiPin-BNC/RCA (cinch), IP67 waterproof 1.8 m Each
15539651 MultiPin-DIN 19262/1 x 4mm banana 1.2 m Each
15549651 MultiPin-Lemo 00/2 x 4mm banana 1.2 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15503181 Supplier Mettler Toledo™ Supplier No. 30281896
  • ONLINE EXCLUSIVE PRICE
    1740.70 NOK
    1615.97 NOK / Pack of 1
    Save 124.73 NOK
    7% Off
In Stock
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter with compatible connector.
  • Detachable
  • Resuable
TRUSTED_SUSTAINABILITY
Specifications
Connector Types MultiPin-BNC/RCA (cinch)
For Use With (Equipment) Mettler Toledo InLab Sensors
For Use With (Application) To connect Mettler Toledo InLab sensors to any meter
Length (English) Cable 4 ft.
Length (Metric) Cable 1.2 m